missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone

CAS: 1070-70-8 | C10H14O4 | 198.22 g/mol

Supplier:  Thermo Scientific Chemicals 03278022

 View more versions of this product

Catalog No. AA3278022

Only null left
Explore available promotions

Description

Description

1,4-Butanediol diacrylate is used as a multifunctional crosslinker. It is used in drying technology for paintings, coatings, inks, resins and adhesives. It serves as an intermediate product for polymer synthesis in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,4-Butanediol diacrylate
1070-70-8
1.051 g/mL
>110°C (230°F)
(Reactive Esters)
1.456
MFCD00014940
UN1760
1,4-butanediol diacrylate, butylene diacrylate, butanediol diacrylate, tetramethylene glycol diacrylate, 1,4-butylene glycol diacrylate, tetramethylene acrylate, tetramethylene diacrylate, 1,4-butylene diacrylate, 1,4-butanediyl diacrylate, 1,4-butanediyl bisacrylate
JHWGFJBTMHEZME-UHFFFAOYSA-N
4-prop-2-enoyloxybutyl prop-2-enoate
70613
≥85%
Stabilized with 50 to 105 ppm hydroquinone
Colorless
83°C (0.3 mmHg)
Pleasant
C10H14O4
CH2=CHCO2(CH2)4O2CCH=CH2
100 g
1777487
Immiscible with water.
C=CC(=O)OCCCCOC(=O)C=C
198.22
198.22
Liquid

Chemical Identifiers

1070-70-8
198.22
JHWGFJBTMHEZME-UHFFFAOYSA-N
70613
C10H14O4
MFCD00014940
1,4-butanediol diacrylate, butylene diacrylate, butanediol diacrylate, tetramethylene glycol diacrylate, 1,4-butylene glycol diacrylate, tetramethylene acrylate, tetramethylene diacrylate, 1,4-butylene diacrylate, 1,4-butanediyl diacrylate, 1,4-butanediyl bisacrylate
C=CC(=O)OCCCCOC(=O)C=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.

P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H312-H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.

EINECSNumber : 213-979-6

RTECSNumber : UD3130000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only