missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone
Description
1,4-Butanediol diacrylate is used as a multifunctional crosslinker. It is used in drying technology for paintings, coatings, inks, resins and adhesives. It serves as an intermediate product for polymer synthesis in the chemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Density | 1.051 g/mL |
| Boiling Point | 83°C (0.3 mmHg) |
| Flash Point | >110°C (230°F) |
| Odor | Pleasant |
| Refractive Index | 1.456 |
| Linear Formula | CH2=CHCO2(CH2)4O2CCH=CH2 |
| Quantity | 5 g |
| UN Number | UN1760 |
| Beilstein | 1777487 |
| Show More |
Safety and Handling
| Hazard Category | H312-H314-H317-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312-H317 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. May cause an allergic skin reaction. |
| Precautionary Statement | P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S. |
| EINECSNumber | 213-979-6 |
| RTECSNumber | UD3130000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only