accessibility menu, dialog, popup

UserName

CAS RN 1070-70-8

IUPAC Name: 4-(prop-2-enoyloxy)butyl prop-2-enoate
Synonyms: butanediol diacrylate
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: JHWGFJBTMHEZME-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCCCOC(=O)C=C
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)