missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)ethylamine 98.0+%, TCI America™

Catalog No. p-7119588
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22665G 5 g
B226625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22665G Supplier TCI America Supplier No. B22665G
Only null left

Chemical Identifiers

CAS 24358-62-1
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00025548
InChI Key SOZMSEPDYJGBEK-UHFFFAOYSA-N
Synonym 1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
PubChem CID 91175
IUPAC Name 1-(4-bromophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Br)N

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 200.08
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(4-Bromophenyl)ethylamine