accessibility menu, dialog, popup

UserName

CAS RN 24358-62-1

IUPAC Name: (1R)-1-(4-bromophenyl)ethan-1-aminium
Synonyms: (1R)-1-(4-bromophenyl)ethanaminium
Molecular Weight (g/mol): 201.09
Molecular Formula: C8H11BrN
InChi Key: SOZMSEPDYJGBEK-ZCFIWIBFSA-O
SMILES: C[C@@H]([NH3+])C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)