missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-(4-Bromophenyl)ethylamine, 96%

Catalog No. AAL1184803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1184803 1 g
AAL1184806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1184803 Supplier Thermo Scientific Chemicals Supplier No. L1184803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24358-62-1 | C8H10BrN | 200.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24358-62-1
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00025548
InChI Key SOZMSEPDYJGBEK-UHFFFAOYSA-N
Synonym 1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
PubChem CID 91175
IUPAC Name 1-(4-bromophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Br)N

Specifications

Density 1.397
Boiling Point 140°C to 145°C (30 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.567
Quantity 1 g
UN Number UN2735
Beilstein 2412318
Sensitivity Air Sensitive
Formula Weight 200.08
Percent Purity 96%
Chemical Name or Material (+/-)-1-(4-Bromophenyl)ethylamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 246-200-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only