missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile, 96%

Catalog No. p-7039851
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59323MD 250 mg
AAH5932303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59323MD Supplier Thermo Scientific Chemicals Supplier No. H59323MD

CAS: 749269-73-6 | C10H7BrFN | 240.075 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 749269-73-6
Molecular Formula C10H7BrFN
Molecular Weight (g/mol) 240.075
MDL Number MFCD19692057
InChI Key KVWJVBSLLYGPIM-UHFFFAOYSA-N
Synonym 1-4-bromo-3-fluorophenyl cyclopropane-1-carbonitrile, 1-4-bromo-3-fluorophenyl cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-4-bromo-3-fluorophenyl, acmc-209owe, 4-bromo-3-fluorophenylcyclopropanenitrile
PubChem CID 11715667
IUPAC Name 1-(4-bromo-3-fluorophenyl)cyclopropane-1-carbonitrile
SMILES C1CC1(C#N)C2=CC(=C(C=C2)Br)F

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 240.07
Percent Purity 96%
Chemical Name or Material 1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only