CAS RN 749269-73-6
IUPAC Name:
1-(4-bromo-3-fluorophenyl)cyclopropane-1-carbonitrile
Synonyms:
1-(4-bromo-3-fluorophenyl)cyclopropane-1-carbonitrile
Molecular Weight (g/mol):
240.08
Molecular Formula:
C10H7BrFN
InChi Key:
KVWJVBSLLYGPIM-UHFFFAOYSA-N
SMILES:
FC1=C(Br)C=CC(=C1)C1(CC1)C#N
1
–
1
of
1
results
1
Promotions Available
1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile, 96%
1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile, 96%
CAS: 749269-73-6 Molecular Formula: C10H7BrFN Molecular Weight (g/mol): 240.075 MDL Number: MFCD19692057 InChI Key: KVWJVBSLLYGPIM-UHFFFAOYSA-N Synonym: 1-4-bromo-3-fluorophenyl cyclopropane-1-carbonitrile,1-4-bromo-3-fluorophenyl cyclopropanecarbonitrile,cyclopropanecarbonitrile, 1-4-bromo-3-fluorophenyl,acmc-209owe,4-bromo-3-fluorophenylcyclopropanenitrile PubChem CID: 11715667 IUPAC Name: 1-(4-bromo-3-fluorophenyl)cyclopropane-1-carbonitrile SMILES: C1CC1(C#N)C2=CC(=C(C=C2)Br)F