missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trifluoromethyl)benzene 99.0+%, TCI America™

Catalog No. p-7119038
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
B140825G 25 g
B1408500G 500 g
2 options

Catalog No. B140825G

Supplier: TCI America B140825G

Only null left

Chemical Identifiers

CAS 433-19-2
Molecular Formula C8H4F6
Molecular Weight (g/mol) 214.11
MDL Number MFCD00000402
InChI Key PDCBZHHORLHNCZ-UHFFFAOYSA-N
Synonym 1,4-bis trifluoromethyl benzene, 1,4-di trifluoromethyl benzene, 1,4-bis trifluoromethyl-benzene, hexafluoro-p-xylene, p-bis trifluoromethyl benzene, benzene, 1,4-bis trifluoromethyl, 1,4-bis-trifluoromethylbenzene, p-trifluoromethylbenzotrifluoride, ptf-tfm, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-p-xylene
PubChem CID 67945
IUPAC Name 1,4-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=C(C=C1)C(F)(F)F

Specifications

Color Colorless
Boiling Point 117°C
Quantity 25 g
UN Number 1993
Formula Weight 214.11
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Bis(trifluoromethyl)benzene
EINECSNumber (3)-3850
TSCA Yes