accessibility menu, dialog, popup

UserName

FFFFFF

CAS RN 433-19-2

IUPAC Name: 1,4-bis(trifluoromethyl)benzene
Synonyms: 1,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 214.11
Molecular Formula: C8H4F6
InChi Key: PDCBZHHORLHNCZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)