missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trifluorométhyl)benzène 99,0+%, TCI America™

Catalog No. B1408500G
Change view
Click to view available options
Quantité:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B1408500G 500 g
B140825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1408500G Supplier TCI America Supplier No. B1408500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 433-19-2
Formule moléculaire C8H4F6
Poids moléculaire (g/mol) 214.11
Numéro MDL MFCD00000402
Clé InChI PDCBZHHORLHNCZ-UHFFFAOYSA-N
Synonyme 1,4-bis trifluoromethyl benzene, 1,4-di trifluoromethyl benzene, 1,4-bis trifluoromethyl-benzene, hexafluoro-p-xylene, p-bis trifluoromethyl benzene, benzene, 1,4-bis trifluoromethyl, 1,4-bis-trifluoromethylbenzene, p-trifluoromethylbenzotrifluoride, ptf-tfm, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-p-xylene
PubChem CID 67945
Nom de l’IUPAC 1,4-bis(trifluorométhyl)benzène
SOURIRES FC(F)(F)C1=CC=C(C=C1)C(F)(F)F

Specifications

Nom ou substance chimique 1,4-Bis(trifluoromethyl)benzene
Couleur Incolore
Point d’ébullition 117 °C
Quantité 500 g
Numéro UN 1993
Poids de formule 214.11
Pourcentage de pureté ≥99,0% (GC)
Forme physique Liquide
EINECSNumber (3)-3850
TSCA Oui