missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(methoxymethyl)benzene 99.0+%, TCI America™
$54.06 - $316.89
Chemical Identifiers
| CAS | 6770-38-3 |
|---|---|
| Molecular Formula | C10H14O2 |
| Molecular Weight (g/mol) | 166.22 |
| MDL Number | MFCD00025903 |
| InChI Key | DAJPMKAQEUGECW-UHFFFAOYSA-N |
| Synonym | p-Xylene Glycol Dimethyl Ether |
| PubChem CID | 81239 |
| IUPAC Name | 1,4-bis(methoxymethyl)benzene |
| SMILES | COCC1=CC=C(C=C1)COC |
Chemical Identifiers
| 6770-38-3 | |
| 166.22 | |
| DAJPMKAQEUGECW-UHFFFAOYSA-N | |
| 81239 | |
| COCC1=CC=C(C=C1)COC |
| C10H14O2 | |
| MFCD00025903 | |
| p-Xylene Glycol Dimethyl Ether | |
| 1,4-bis(methoxymethyl)benzene |
Specifications
| 6770-38-3 | |
| 235°C | |
| MFCD00025903 | |
| p-Xylene Glycol Dimethyl Ether | |
| COCC1=CC=C(C=C1)COC | |
| 166.22 | |
| 166.22 | |
| Liquid |
| Colorless | |
| C10H14O2 | |
| 25 g | |
| DAJPMKAQEUGECW-UHFFFAOYSA-N | |
| 1,4-bis(methoxymethyl)benzene | |
| 81239 | |
| ≥99.0% (GC) | |
| 1,4-Bis(methoxymethyl)benzene |
Safety and Handling
EINECSNumber : (3)-2883
TSCA : Yes