accessibility menu, dialog, popup

UserName

CAS RN 6770-38-3

IUPAC Name: 1,4-bis(methoxymethyl)benzene
Synonyms: 1,4-bis(methoxymethyl)benzene
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: DAJPMKAQEUGECW-UHFFFAOYSA-N
SMILES: COCC1=CC=C(COC)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results