missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(methoxymethyl)benzene 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 235°C |
| Quantity | 500 g |
| Formula Weight | 166.22 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,4-Bis(methoxymethyl)benzene |
Safety and Handling
| EINECSNumber | (3)-2883 |
| TSCA | Yes |