missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(bromomethyl)naphthalene (contains isomer) 65.0+%, TCI America™

Catalog No. b19195g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19195G 5 g
B19191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19195G Supplier TCI America Supplier No. B19195G

Chemical Identifiers

CAS 58791-49-4
Molecular Formula C12H10Br2
Molecular Weight (g/mol) 314.02
MDL Number MFCD01321143
InChI Key UZHZQZOMHXNQBJ-UHFFFAOYSA-N
PubChem CID 593017
IUPAC Name 1,4-bis(bromomethyl)naphthalene
SMILES C1=CC=C2C(=CC=C(C2=C1)CBr)CBr

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 314.02
Percent Purity ≥65.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(bromomethyl)naphthalene (contains isomer)