accessibility menu, dialog, popup

UserName

BrBr

CAS RN 58791-49-4

IUPAC Name: 1,4-bis(bromomethyl)naphthalene
Synonyms: 1,4-bis(bromomethyl)naphthalene
Molecular Weight (g/mol): 314.02
Molecular Formula: C12H10Br2
InChi Key: UZHZQZOMHXNQBJ-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(CBr)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results