missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(bromomethyl)-2,5-dimethylbenzene 98.0+%, TCI America™

Catalog No. B22105G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22105G 5 g
B22101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22105G Supplier TCI America Supplier No. B22105G

Chemical Identifiers

CAS 35168-62-8
Molecular Formula C10H12Br2
Molecular Weight (g/mol) 292.014
MDL Number MFCD04038409
InChI Key MUSYLRHTIZVVCB-UHFFFAOYSA-N
Synonym 2,5-Bis(bromomethyl)-p-xylene, 2,5-Dimethyl-1,4-bis(bromomethyl)benzene
PubChem CID 3015742
IUPAC Name 1,4-bis(bromomethyl)-2,5-dimethylbenzene
SMILES CC1=CC(=C(C=C1CBr)C)CBr

Specifications

Melting Point 163°C
Quantity 5 g
UN Number 1759
Formula Weight 292.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(bromomethyl)-2,5-dimethylbenzene