CAS RN 35168-62-8
IUPAC Name:
1,4-bis(bromomethyl)-2,5-dimethylbenzene
Synonyms:
1,4-bis(bromomethyl)-2,5-dimethylbenzene
Molecular Weight (g/mol):
292.01
Molecular Formula:
C10H12Br2
InChi Key:
MUSYLRHTIZVVCB-UHFFFAOYSA-N
SMILES:
CC1=CC(CBr)=C(C)C=C1CBr
1
–
1
of
1
results
1
1,4-Bis(bromomethyl)-2,5-dimethylbenzene 98.0+%, TCI America™
CAS: 35168-62-8 Molecular Formula: C10H12Br2 Molecular Weight (g/mol): 292.014 MDL Number: MFCD04038409 InChI Key: MUSYLRHTIZVVCB-UHFFFAOYSA-N Synonym: 2,5-Bis(bromomethyl)-p-xylene, 2,5-Dimethyl-1,4-bis(bromomethyl)benzene PubChem CID: 3015742 IUPAC Name: 1,4-bis(bromomethyl)-2,5-dimethylbenzene SMILES: CC1=CC(=C(C=C1CBr)C)CBr