missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(2-hydroxyisopropyl)benzene, 97%

Catalog No. AAB2147203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2147203 1 g
AAB2147206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2147203 Supplier Thermo Scientific Chemicals Supplier No. B2147203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2948-46-1 | C12H18O2 | 194.274 g/mol

1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.

Solubility
Slightly Soluble in water (2.1 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.

Chemical Identifiers

CAS 2948-46-1
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.274
MDL Number MFCD00009827
InChI Key LEARFTRDZQQTDN-UHFFFAOYSA-N
Synonym 1,4-bis 2-hydroxy-2-propyl benzene, 1,4-bis 1-methyl-1-hydroxyethyl benzene, alpha,alpha,alpha',alpha'-tetramethyl-1,4-benzenedimethanol, unii-g785drb5xh, p-bis 2-hydroxyisopropyl benzene, 1,4-bis 2-hydroxyisopropyl benzene, p-bis alpha-hydroxyisopropyl benzene, g785drb5xh, alpha,alpha'-dihydroxy-p-diisopropylbenzene, alpha,alpha,alpha',alpha'-tetramethyl-p-xylylenediol
PubChem CID 18062
IUPAC Name 2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES CC(C)(C1=CC=C(C=C1)C(C)(C)O)O

Specifications

Melting Point 142°C to 146°C
Boiling Point 167°C to 168°C (12 mmHg)
Quantity 1 g
Solubility Information Slightly Soluble in water (2.1g/L) (25°C).
Formula Weight 194.28
Percent Purity 97%
Chemical Name or Material 1,4-Bis(2-hydroxyisopropyl)benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-964-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only