Learn More
1,4-Benzenedithiol, 97%
CAS: 624-39-5 | C6H6S2 | 142.23 g/mol
Supplier: Thermo Scientific Chemicals L19440MD
Description
1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications
| 1,4-Benzenedithiol | |
| 95°C to 99°C | |
| C6H6S2 | |
| 250 mg | |
| 1680022 | |
| 1,4-benzenedithiol, dithiohydroquinone, 1,4-dimercaptobenzene, 4-mercaptothiophenol, 1,4-benzene dithiol, pubchem6866, acmc-1b45q, benzene-1,4-dithiol gc | |
| WYLQRHZSKIDFEP-UHFFFAOYSA-N | |
| benzene-1,4-dithiol | |
| 4691729 | |
| 97% |
| 624-39-5 | |
| Stench | |
| MFCD00142541 | |
| UN3335 | |
| Air sensitive | |
| Soluble in water. | |
| SC1=CC=C(S)C=C1 | |
| 142.23 | |
| 142.24 |
Chemical Identifiers
| 624-39-5 | |
| 142.23 | |
| WYLQRHZSKIDFEP-UHFFFAOYSA-N | |
| 4691729 | |
| SC1=CC=C(S)C=C1 |
| C6H6S2 | |
| MFCD00142541 | |
| 1,4-benzenedithiol, dithiohydroquinone, 1,4-dimercaptobenzene, 4-mercaptothiophenol, 1,4-benzene dithiol, pubchem6866, acmc-1b45q, benzene-1,4-dithiol gc | |
| benzene-1,4-dithiol |
Safety and Handling
GHS H Statement
H318-H302-H312-H332-H335-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H318-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only