missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedithiol, 97%

Catalog No. p-7047555
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
5 g
Catalog No. Quantity
AAL19440MD 250 mg
AAL1944003 1 g
AAL1944006 5 g
3 options

Catalog No. AAL19440MD

Supplier: Thermo Scientific Chemicals L19440MD

Only null left

CAS: 624-39-5 | C6H6S2 | 142.23 g/mol

1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 624-39-5
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00142541
InChI Key WYLQRHZSKIDFEP-UHFFFAOYSA-N
Synonym 1,4-benzenedithiol, dithiohydroquinone, 1,4-dimercaptobenzene, 4-mercaptothiophenol, 1,4-benzene dithiol, pubchem6866, acmc-1b45q, benzene-1,4-dithiol gc
PubChem CID 4691729
IUPAC Name benzene-1,4-dithiol
SMILES SC1=CC=C(S)C=C1

Specifications

Melting Point 95°C to 99°C
Odor Stench
Quantity 250 mg
UN Number UN3335
Beilstein 1680022
Sensitivity Air sensitive
Solubility Information Soluble in water.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 1,4-Benzenedithiol
Hazard Category H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H312-H332-H335-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only