missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenylurea 98.0+%, TCI America™
Supplier: TCI America C0031500G
Specifications
| 1,3-Diphenylurea | |
| 240°C | |
| 260°C | |
| MFCD00003017 | |
| n,n'-diphenylurea, carbanilide, diphenylurea, diphenylcarbamide, s-diphenylurea, urea, n,n'-diphenyl, sym-diphenylurea, acardite i, acardite, n-phenyl-n'-phenylurea | |
| O=C(NC1=CC=CC=C1)NC1=CC=CC=C1 | |
| 212.25 | |
| CHEBI:41320 | |
| ≥98.0% (HPLC,N) |
| 102-07-8 | |
| White | |
| C13H12N2O | |
| 500 g | |
| GWEHVDNNLFDJLR-UHFFFAOYSA-N | |
| 1,3-diphenylurea | |
| 7595 | |
| 212.25 | |
| Crystalline Powder |
Chemical Identifiers
| 102-07-8 | |
| 212.25 | |
| GWEHVDNNLFDJLR-UHFFFAOYSA-N | |
| 7595 | |
| 1,3-diphenylurea |
| C13H12N2O | |
| MFCD00003017 | |
| n,n'-diphenylurea, carbanilide, diphenylurea, diphenylcarbamide, s-diphenylurea, urea, n,n'-diphenyl, sym-diphenylurea, acardite i, acardite, n-phenyl-n'-phenylurea | |
| CHEBI:41320 | |
| O=C(NC1=CC=CC=C1)NC1=CC=CC=C1 |
Safety and Handling
RTECSNumber : FD9800000
TSCA : Yes