accessibility menu, dialog, popup

UserName

OHNHN

CAS RN 102-07-8

IUPAC Name: 1,3-diphenylurea
Synonyms: 1,3-diphenylurea N,N'-difenylmocovina diphenyl urea, unsym usaf EK-534 acardite I diphenylurea, sym acardite karbanilid
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: GWEHVDNNLFDJLR-UHFFFAOYSA-N
SMILES: O=C(NC1=CC=CC=C1)NC1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)