missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenylurea 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 240°C |
| Color | White |
| Boiling Point | 260°C |
| Quantity | 25 g |
| Formula Weight | 212.25 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3-Diphenylurea |
Safety and Handling
| RTECSNumber | FD9800000 |
| TSCA | Yes |