missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenyltetramethyldisilazane 95.0+%, TCI America™
Supplier: TCI America D181625ML
Specifications
| 1,3-Diphenyltetramethyldisilazane | |
| Yellow | |
| C16H23NSi2 | |
| 25 mL | |
| HIMXYMYMHUAZLW-UHFFFAOYSA-N | |
| bis[dimethyl(phenyl)silyl]amine | |
| 76988 | |
| ≥95.0% (GC) |
| 3449-26-1 | |
| 146°C | |
| MFCD00042916 | |
| Tetramethyl-1,3-diphenyldisilazane | |
| C[Si](C)(N[Si](C)(C)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 285.54 | |
| 285.54 | |
| Liquid |
Chemical Identifiers
| 3449-26-1 | |
| 285.54 | |
| HIMXYMYMHUAZLW-UHFFFAOYSA-N | |
| 76988 | |
| C[Si](C)(N[Si](C)(C)C1=CC=CC=C1)C1=CC=CC=C1 |
| C16H23NSi2 | |
| MFCD00042916 | |
| Tetramethyl-1,3-diphenyldisilazane | |
| bis[dimethyl(phenyl)silyl]amine |
Safety and Handling
TSCA : No