accessibility menu, dialog, popup

UserName

CH3SiH3CHNSiCH3H3C

CAS RN 3449-26-1

IUPAC Name: bis[dimethyl(phenyl)silyl]amine
Synonyms: bis[dimethyl(phenyl)silyl]amine
Molecular Weight (g/mol): 285.54
Molecular Formula: C16H23NSi2
InChi Key: HIMXYMYMHUAZLW-UHFFFAOYSA-N
SMILES: C[Si](C)(N[Si](C)(C)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results