missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyltetramethyldisilazane 95.0+%, TCI America™

Catalog No. p-7126947
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18165ML 5 mL
D181625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18165ML Supplier TCI America Supplier No. D18165ML
Only null left

Chemical Identifiers

CAS 3449-26-1
Molecular Formula C16H23NSi2
Molecular Weight (g/mol) 285.54
MDL Number MFCD00042916
InChI Key HIMXYMYMHUAZLW-UHFFFAOYSA-N
Synonym Tetramethyl-1,3-diphenyldisilazane
PubChem CID 76988
IUPAC Name bis[dimethyl(phenyl)silyl]amine
SMILES C[Si](C)(N[Si](C)(C)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 mL
Formula Weight 285.54
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diphenyltetramethyldisilazane