missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99%, Spectrum™ Chemical

Catalog No. 18608059
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608059 4 L Steel Can
18608060 500 mL Steel Can
18608057 20 L Steel Can
18608058 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Catalog No. 18608059 missing translation for 'mfr' Spectrum Chemical Mfg Cor missing translation for 'supplierNo' D23714LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 646-06-0

Specifications

Acidity 100 ppm
CAS Max % 1
Boiling Point 74 to 78°C
Packaging Steel Can
Quantity 4 L
Percent Purity 0.99
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.