missing translation for 'onlineSavingsMsg'
Learn More

(1,3-Dioxolan-2-ylmethyl)magnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC438861000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438861000 100 mL AcroSeal™ Glass Bottle
AC438868000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438861000 Supplier Thermo Scientific Chemicals Supplier No. 438861000

CAS: 180675-22-3 | C4H7BrMgO2 | 191.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 180675-22-3 , 109-99-9
Molecular Formula C4H7BrMgO2
Molecular Weight (g/mol) 191.31
MDL Number MFCD01321271
InChI Key ZDNTZRAFJZERSL-UHFFFAOYSA-M
Synonym 1,3-dioxolan-2-ylmethyl magnesium bromide solution, magnesium 2-methanidyl-1,3-dioxolane bromide, magnesium 2+ ion 1,3-dioxolan-2-ylmethanide bromide, grignard reagent
PubChem CID 4247254
IUPAC Name magnesium;2-methanidyl-1,3-dioxolane;bromide
SMILES Br[Mg]CC1OCCO1

Specifications

CAS Min % 8.14, 87.79
CAS Max % 12.21, 91.86
Color Brown to White
Density 0.9400g/mL
Boiling Point 65.0°C to 67.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.94
Formula Weight 191.31
Physical Form Powder
Chemical Name or Material (1, 3-Dioxolan-2-ylmethyl)magnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only