missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1,3-Dioxolan-2-yl)methyltriphenylphosphonium Bromide 97.0+%, TCI America™
$70.84 - $242.33
Chemical Identifiers
| CAS | 52509-14-5 |
|---|---|
| Molecular Formula | C22H22BrO2P |
| Molecular Weight (g/mol) | 429.29 |
| MDL Number | MFCD00011966 |
| InChI Key | FRHRVQQUICVJDG-UHFFFAOYSA-M |
| Synonym | 1,3-dioxolan-2-ylmethyl triphenylphosphonium bromide, 1,3-dioxolan-2-yl methyl triphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphanium bromide, phosphonium, 1,3-dioxolan-2-ylmethyl triphenyl-, bromide, 1,3-dioxolan-2-yl methyltriphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphanium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphine, bromide, acmc-209kzy |
| PubChem CID | 2724194 |
| IUPAC Name | [(1,3-dioxolan-2-yl)methyl]triphenylphosphanium bromide |
| SMILES | [Br-].C(C1OCCO1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 52509-14-5 | |
| 429.29 | |
| FRHRVQQUICVJDG-UHFFFAOYSA-M | |
| 2724194 | |
| [Br-].C(C1OCCO1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C22H22BrO2P | |
| MFCD00011966 | |
| 1,3-dioxolan-2-ylmethyl triphenylphosphonium bromide, 1,3-dioxolan-2-yl methyl triphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphanium bromide, phosphonium, 1,3-dioxolan-2-ylmethyl triphenyl-, bromide, 1,3-dioxolan-2-yl methyltriphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphanium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphine, bromide, acmc-209kzy | |
| [(1,3-dioxolan-2-yl)methyl]triphenylphosphanium bromide |
Specifications
| 52509-14-5 | |
| C22H22BrO2P | |
| 5 g | |
| FRHRVQQUICVJDG-UHFFFAOYSA-M | |
| [(1,3-dioxolan-2-yl)methyl]triphenylphosphanium bromide | |
| 2724194 | |
| ≥97.0% (T) | |
| (1,3-Dioxolan-2-yl)methyltriphenylphosphonium Bromide |
| 191°C | |
| MFCD00011966 | |
| 1,3-dioxolan-2-ylmethyl triphenylphosphonium bromide, 1,3-dioxolan-2-yl methyl triphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphanium bromide, phosphonium, 1,3-dioxolan-2-ylmethyl triphenyl-, bromide, 1,3-dioxolan-2-yl methyltriphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphanium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphine, bromide, acmc-209kzy | |
| [Br-].C(C1OCCO1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 429.29 | |
| 429.29 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6734
TSCA : No