missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diisopropylthiourea 99.0+%, TCI America™
Supplier: TCI America D025325G
Specifications
| 1,3-Diisopropylthiourea | |
| 144°C | |
| C7H16N2S | |
| 25 g | |
| CC(C)NC(=S)NC(C)C | |
| 160.28 | |
| 160.28 | |
| Crystalline Powder |
| 2986-17-6 | |
| White-Yellow | |
| MFCD00040485 | |
| KREOCUNMMFZOOS-UHFFFAOYSA-N | |
| 1,3-bis(propan-2-yl)thiourea | |
| 2758386 | |
| ≥99.0% (N) |
Chemical Identifiers
| 2986-17-6 | |
| 160.28 | |
| KREOCUNMMFZOOS-UHFFFAOYSA-N | |
| 1,3-bis(propan-2-yl)thiourea |
| C7H16N2S | |
| MFCD00040485 | |
| 2758386 | |
| CC(C)NC(=S)NC(C)C |
Safety and Handling
EINECSNumber : (2)-1757&(2)-1759
RTECSNumber : YS9860000
TSCA : Yes