accessibility menu, dialog, popup

UserName

CAS RN 2986-17-6

H3CCH3HNSHNCH3CH3

CAS RN 2986-17-6

IUPAC Name: 1,3-bis(propan-2-yl)thiourea
Synonyms: 1,3-diisopropylthiourea
Molecular Weight (g/mol): 160.28
Molecular Formula: C7H16N2S
InChi Key: KREOCUNMMFZOOS-UHFFFAOYSA-N
SMILES: CC(C)NC(=S)NC(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results