missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodobenzene 99.0+%, TCI America™

Catalog No. p-7126317
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06075G 5 g
D060725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06075G Supplier TCI America Supplier No. D06075G
Only null left

Chemical Identifiers

CAS 626-00-6
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.907
MDL Number MFCD00041731
InChI Key SFPQFQUXAJOWNF-UHFFFAOYSA-N
Synonym m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo
PubChem CID 12270
IUPAC Name 1,3-diiodobenzene
SMILES C1=CC(=CC(=C1)I)I

Specifications

Melting Point 35°C
Boiling Point 284°C
Quantity 5 g
Formula Weight 329.91
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diiodobenzene
TSCA Yes
Recommended Storage Refrigerator