accessibility menu, dialog, popup

UserName

CAS RN 626-00-6

IUPAC Name: 1,3-diiodobenzene
Synonyms: 1,3-diiodobenzene
Molecular Weight (g/mol): 329.91
Molecular Formula: C6H4I2
InChi Key: SFPQFQUXAJOWNF-UHFFFAOYSA-N
SMILES: IC1=CC(I)=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)