Learn More
1,3-Diiodobenzene, 98%
CAS: 626-00-6 | C6H4I2 | 329.9 g/mol
$60.34 - $60.34
Chemical Identifiers
| CAS | 626-00-6 |
|---|---|
| Molecular Formula | C6H4I2 |
| Molecular Weight (g/mol) | 329.9 |
| MDL Number | MFCD00041731 |
| InChI Key | SFPQFQUXAJOWNF-UHFFFAOYSA-N |
| Synonym | m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo |
| PubChem CID | 12270 |
| IUPAC Name | 1,3-diiodobenzene |
| SMILES | C1=CC(=CC(=C1)I)I |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC407740050
|
Thermo Scientific Chemicals
407740050 |
5 g | Glass bottle |
Each for $60.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 626-00-6 | |
| 329.9 | |
| SFPQFQUXAJOWNF-UHFFFAOYSA-N | |
| 12270 | |
| C1=CC(=CC(=C1)I)I |
| C6H4I2 | |
| MFCD00041731 | |
| m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo | |
| 1,3-diiodobenzene |
Specifications
| 626-00-6 | |
| 100.0 | |
| Yellow to Beige | |
| Authentic | |
| Glass bottle | |
| C6H4I2 | |
| 5 g | |
| m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo | |
| SFPQFQUXAJOWNF-UHFFFAOYSA-N | |
| 1,3-diiodobenzene | |
| 12270 | |
| 98% | |
| 1, 3-Diiodobenzene, 98% |
| 97.5 | |
| 33°C to 37°C | |
| 2.4700g/mL | |
| 97.5% min. (GC) | |
| C6H4I2 | |
| MFCD00041731 | |
| 2.47 | |
| Solubility in water: insoluble | |
| C1=CC(=CC(=C1)I)I | |
| 329.9 | |
| 329.9 | |
| Low Melting Crystalline Mass, Powder or Crystals |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
EINECSNumber : 210-921-1
TSCA : TSCA
RUO – Research Use Only