missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodobenzene, 98+%

CAS: 626-00-6 | C6H4I2 | 329.907 g/mol

Supplier:  Thermo Scientific Chemicals L1744514

 View more versions of this product

Catalog No. AAL1744514


Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in hot methanol.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with strong oxidizing agents.
Specifications

Specifications

1,3-Diiodobenzene
35°C to 37°C
285°C
C6H4I2
25 g
Light sensitive
Insoluble in water. Soluble in hot methanol.
C1=CC(=CC(=C1)I)I
329.907
329.91
626-00-6
2.47
>110°C (230°F)
MFCD00041731
1904540
m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo
SFPQFQUXAJOWNF-UHFFFAOYSA-N
1,3-diiodobenzene
12270
≥98%

Chemical Identifiers

626-00-6
329.907
SFPQFQUXAJOWNF-UHFFFAOYSA-N
12270
C1=CC(=CC(=C1)I)I
C6H4I2
MFCD00041731
m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo
1,3-diiodobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-921-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only