missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodobenzene, 98+%

Catalog No. AAL1744506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1744506 5 g
AAL1744514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1744506 Supplier Thermo Scientific Chemicals Supplier No. L1744506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 626-00-6 | C6H4I2 | 329.907 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in hot methanol.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 626-00-6
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.907
MDL Number MFCD00041731
InChI Key SFPQFQUXAJOWNF-UHFFFAOYSA-N
Synonym m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo
PubChem CID 12270
IUPAC Name 1,3-diiodobenzene
SMILES C1=CC(=CC(=C1)I)I

Specifications

Melting Point 35°C to 37°C
Density 2.47
Boiling Point 285°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1904540
Sensitivity Light sensitive
Solubility Information Insoluble in water. Soluble in hot methanol.
Formula Weight 329.91
Percent Purity ≥98%
Chemical Name or Material 1,3-Diiodobenzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-921-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only