missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dihydroxynaphthalene 98.0+%, TCI America™
Supplier: TCI America D05885G
Specifications
| 1,3-Dihydroxynaphthalene | |
| 125°C | |
| MFCD00003965 | |
| 1,3-naphthalenediol, 1,3-dihydroxynaphthalene, naphthoresorcinol, naphthoresoucinol, 3-hydroxybenzocyclohexadien-1-one, naphthoresorcin, ccris 7896, unii-5x457yew8y, acmc-1bvwb, naphthalenediol-1,3 | |
| C1=CC=C2C(=C1)C=C(C=C2O)O | |
| 160.172 | |
| 160.17 | |
| Crystalline Powder |
| 132-86-5 | |
| C10H8O2 | |
| 5 g | |
| XOOMNEFVDUTJPP-UHFFFAOYSA-N | |
| naphthalene-1,3-diol | |
| 8601 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 132-86-5 | |
| 160.172 | |
| XOOMNEFVDUTJPP-UHFFFAOYSA-N | |
| 8601 | |
| C1=CC=C2C(=C1)C=C(C=C2O)O |
| C10H8O2 | |
| MFCD00003965 | |
| 1,3-naphthalenediol, 1,3-dihydroxynaphthalene, naphthoresorcinol, naphthoresoucinol, 3-hydroxybenzocyclohexadien-1-one, naphthoresorcin, ccris 7896, unii-5x457yew8y, acmc-1bvwb, naphthalenediol-1,3 | |
| naphthalene-1,3-diol |
Safety and Handling
EINECSNumber : (4)-0356
RTECSNumber : QJ4725000
TSCA : Yes