missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxynaphthalene, 99+%

Catalog No. p-4510401
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163340010 1 g Glass bottle
AC163340050 5 g Glass bottle
AC163340250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163340010 Supplier Thermo Scientific Chemicals Supplier No. 163340010
Only null left

CAS: 132-86-5 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132-86-5
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
InChI Key XOOMNEFVDUTJPP-UHFFFAOYSA-N
Synonym 1,3-naphthalenediol, 1,3-dihydroxynaphthalene, naphthoresorcinol, naphthoresoucinol, 3-hydroxybenzocyclohexadien-1-one, naphthoresorcin, ccris 7896, unii-5x457yew8y, acmc-1bvwb, naphthalenediol-1,3
PubChem CID 8601
IUPAC Name naphthalene-1,3-diol
SMILES C1=CC=C2C(=C1)C=C(C=C2O)O

Specifications

Melting Point 123.0°C to 126.0°C
Color Brown to Orange-Pink
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C10H6(OH)2
Quantity 1 g
Beilstein 06,978
Merck Index 15,6481
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether,soluble in ethanol an diethyl ether,slightly soluble in acetone,benzene and ligroin
Formula Weight 160.17
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Dihydroxynaphthalene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 205-079-7

RUO – Research Use Only