Learn More
1,3-Dihydroxynaphthalene, 98%
CAS: 132-86-5 | C10H8O2 | 160.172 g/mol
Supplier: Thermo Scientific Chemicals A1773903
Description
It is used in the naphthoresorcinol test for uronic acids of Tollens and Rorive, a blue color can be extracted with ethanol after heating of uronic acids with naphthoresorcinol in hydrochloric acid. It is also employed as a reagent for various medical tests. It can also be used as a spot test reagent for glycerate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the naphthoresorcinol test for uronic acids of Tollens and Rorive, a blue color can be extracted with ethanol after heating of uronic acids with naphthoresorcinol in hydrochloric acid. It is also employed as a reagent for various medical tests. It can also be used as a spot test reagent for glycerate.
Solubility
Soluble in ethanol (50 mg/ml), and water.
Notes
Light Sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry and well ventilated place.
Specifications
| 1,3-Dihydroxynaphthalene | |
| 123°C to 126°C | |
| MFCD00003965 | |
| 2044002 | |
| 14,6396 | |
| Soluble in ethanol (50mg/ml),and water. | |
| C1=CC=C2C(=C1)C=C(C=C2O)O | |
| 160.172 | |
| 160.17 |
| 132-86-5 | |
| C10H8O2 | |
| 1 g | |
| Light sensitive | |
| 1,3-naphthalenediol, 1,3-dihydroxynaphthalene, naphthoresorcinol, naphthoresoucinol, 3-hydroxybenzocyclohexadien-1-one, naphthoresorcin, ccris 7896, unii-5x457yew8y, acmc-1bvwb, naphthalenediol-1,3 | |
| XOOMNEFVDUTJPP-UHFFFAOYSA-N | |
| naphthalene-1,3-diol | |
| 8601 | |
| 98% |
Chemical Identifiers
| 132-86-5 | |
| MFCD00003965 | |
| 1,3-naphthalenediol, 1,3-dihydroxynaphthalene, naphthoresorcinol, naphthoresoucinol, 3-hydroxybenzocyclohexadien-1-one, naphthoresorcin, ccris 7896, unii-5x457yew8y, acmc-1bvwb, naphthalenediol-1,3 | |
| naphthalene-1,3-diol |
| C10H8O2 | |
| XOOMNEFVDUTJPP-UHFFFAOYSA-N | |
| 8601 |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 205-079-7
RTECSNumber : QJ4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only