missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluorobenzene 98.0+%, TCI America™

Catalog No. p-7126832
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D162625G 25 g
D1626500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D162625G Supplier TCI America Supplier No. D162625G
Only null left

Chemical Identifiers

CAS 372-18-9
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000327
InChI Key UEMGWPRHOOEKTA-UHFFFAOYSA-N
Synonym m-difluorobenzene, benzene, 1,3-difluoro, benzene, m-difluoro, 2,4-difluorobenzene, meta-difluorobenzene, 1,3-difluoro benzene, unii-vo3c26woc2, 1,3-difluorbenzol, vo3c26woc2, 1,3difluorobenzene
PubChem CID 9741
ChEBI CHEBI:38584
IUPAC Name 1,3-difluorobenzene
SMILES FC1=CC(F)=CC=C1

Specifications

Melting Point -59°C
Color Yellow
Boiling Point 83°C
Quantity 25 g
UN Number 1993
Formula Weight 114.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Difluorobenzene
EINECSNumber (3)-2688
RTECSNumber CZ5652000
TSCA No