accessibility menu, dialog, popup

UserName

FF

CAS RN 372-18-9

IUPAC Name: 1,3-difluorobenzene
Synonyms: 1,3-difluorobenzene M-difluorobenzene
Molecular Weight (g/mol): 114.1
Molecular Formula: C6H4F2
InChi Key: UEMGWPRHOOEKTA-UHFFFAOYSA-N
SMILES: FC1=CC(F)=CC=C1
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)