Learn More
1,3-Difluorobenzene, 99%
CAS: 372-18-9 | C6H4F2 | 114.10 g/mol
Supplier: Thermo Scientific Chemicals A1438718
Description
It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.
Solubility
Soluble in water at 20°C 1.1g/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 1,3-Difluorobenzene | |
| -59°C | |
| 82°C to 83°C | |
| Characteristic | |
| 1.438 | |
| 50 g | |
| 1904537 | |
| Soluble in water at 20°C 1.1g/L. | |
| FC1=CC(F)=CC=C1 | |
| 114.10 | |
| CHEBI:38584 | |
| 99% |
| 372-18-9 | |
| 1.16 | |
| 2°C (36°F) | |
| C6H4F2 | |
| MFCD00000327 | |
| UN1993 | |
| m-difluorobenzene, benzene, 1,3-difluoro, benzene, m-difluoro, 2,4-difluorobenzene, meta-difluorobenzene, 1,3-difluoro benzene, unii-vo3c26woc2, 1,3-difluorbenzol, vo3c26woc2, 1,3difluorobenzene | |
| UEMGWPRHOOEKTA-UHFFFAOYSA-N | |
| 1,3-difluorobenzene | |
| 9741 | |
| 114.09 |
Chemical Identifiers
| 372-18-9 | |
| 114.10 | |
| UEMGWPRHOOEKTA-UHFFFAOYSA-N | |
| 9741 | |
| 1,3-difluorobenzene |
| C6H4F2 | |
| MFCD00000327 | |
| m-difluorobenzene, benzene, 1,3-difluoro, benzene, m-difluoro, 2,4-difluorobenzene, meta-difluorobenzene, 1,3-difluoro benzene, unii-vo3c26woc2, 1,3-difluorbenzol, vo3c26woc2, 1,3difluorobenzene | |
| CHEBI:38584 | |
| FC1=CC(F)=CC=C1 |
Safety and Handling
GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H225-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 206-746-5
RTECSNumber : CZ5652000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only