missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluorobenzene, 99+%

Catalog No. AC114600100
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114600100 10 mL Glass bottle
AC114600250 25 mL Glass bottle
AC114601000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114600100 Supplier Thermo Scientific Chemicals Supplier No. 114600100

CAS: 372-18-9 | C6H4F2 | 114.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 372-18-9
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000327
InChI Key UEMGWPRHOOEKTA-UHFFFAOYSA-N
Synonym m-difluorobenzene, benzene, 1,3-difluoro, benzene, m-difluoro, 2,4-difluorobenzene, meta-difluorobenzene, 1,3-difluoro benzene, unii-vo3c26woc2, 1,3-difluorbenzol, vo3c26woc2, 1,3difluorobenzene
PubChem CID 9741
ChEBI CHEBI:38584
IUPAC Name 1,3-difluorobenzene
SMILES FC1=CC(F)=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -59°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 83°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4370 to 1.439
Linear Formula C6H4F2
Quantity 10 mL
Beilstein 05, I, 108
Specific Gravity 1.15
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,acetone,dmf,,2-propanol and methylene chloride
Formula Weight 114.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 3-Difluorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-746-5

RUO – Research Use Only