missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoro-2-nitrobenzene, 98%

Catalog No. p-7037103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5569403 1 g
AAH5569406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5569403 Supplier Thermo Scientific Chemicals Supplier No. H5569403
Only null left

CAS: 19064-24-5 | C6H3F2NO2 | 159.092 g/mol

1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.

Chemical Identifiers

CAS 19064-24-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.092
MDL Number MFCD00192035
InChI Key SSNCMIDZGFCTST-UHFFFAOYSA-N
Synonym 2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen
PubChem CID 87922
IUPAC Name 1,3-difluoro-2-nitrobenzene
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])F

Specifications

Melting Point 0°C
Density 1.503
Boiling Point 91°C to 92°C (11 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.494
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 1,3-Difluoro-2-nitrobenzene
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 242-793-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only