missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethoxybenzene 97.0+%, TCI America™

Catalog No. D045325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D045325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D045325G Supplier TCI America Supplier No. D045325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2049-73-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00015145
InChI Key MKGFYMKFBCWNCP-UHFFFAOYSA-N
Synonym m-diethoxybenzene, benzene, 1,3-diethoxy, resorcinol diethyl ether, resorcindiathylather, benzene,3-diethoxy, acmc-1cdp4, benzene, 1,3-diethoxy-9ci
PubChem CID 74899
IUPAC Name 1,3-diethoxybenzene
SMILES CCOC1=CC(=CC=C1)OCC

Specifications

Color Yellow
Boiling Point 235°C
Quantity 25 g
Formula Weight 166.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diethoxybenzene