accessibility menu, dialog, popup

UserName

CH3OOH3C

CAS RN 2049-73-2

IUPAC Name: 1,3-diethoxybenzene
Synonyms: 1,3-diethoxybenzene
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: MKGFYMKFBCWNCP-UHFFFAOYSA-N
SMILES: CCOC1=CC(OCC)=CC=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)