missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethoxybenzene, 95%

Catalog No. AAA1133809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1133809 10 g
AAA1133818 50 g
AAA1133830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1133809 Supplier Thermo Scientific Chemicals Supplier No. A1133809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2049-73-2 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2049-73-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00015145
InChI Key MKGFYMKFBCWNCP-UHFFFAOYSA-N
Synonym m-diethoxybenzene, benzene, 1,3-diethoxy, resorcinol diethyl ether, resorcindiathylather, benzene,3-diethoxy, acmc-1cdp4, benzene, 1,3-diethoxy-9ci
PubChem CID 74899
IUPAC Name 1,3-diethoxybenzene
SMILES CCOC1=CC(=CC=C1)OCC

Specifications

Melting Point 11°C to 12°C
Density 1.01
Boiling Point 234°C to 236°C
Refractive Index 1.5095
Quantity 10 g
Beilstein 2517730
Formula Weight 166.22
Percent Purity 95%
Chemical Name or Material 1,3-Diethoxybenzene
EINECSNumber 218-071-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only