missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromobutane, 98%

Catalog No. p-7027400
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2412314 25 g
AAB2412322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2412314 Supplier Thermo Scientific Chemicals Supplier No. B2412314

CAS: 107-80-2 | C4H8Br2 | 215.916 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-80-2
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.916
MDL Number MFCD00000152
InChI Key XZNGUVQDFJHPLU-UHFFFAOYSA-N
Synonym butane, 1,3-dibromo, 1,3-butylene bromide, 1,3 dibromobutane, 1,3-dibrombutan, butane,1,3-dibromo, 1,3-dibromo-butan, 1,3-butylenebromide, 1,3-dibromo butane, 1,3-dibromo-butane, pubchem3866
PubChem CID 7889
IUPAC Name 1,3-dibromobutane
SMILES CC(CCBr)Br

Specifications

Density 1.789
Boiling Point 173°C to 176°C
Flash Point None
Refractive Index 1.5095
Quantity 25 g
Beilstein 1731231
Formula Weight 215.93
Percent Purity 98%
Chemical Name or Material 1,3-Dibromobutane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-520-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only