accessibility menu, dialog, popup

UserName

CAS RN 107-80-2

IUPAC Name: 1,3-dibromobutane
Synonyms: butane, 1,3-dibromo-
Molecular Weight (g/mol): 215.92
Molecular Formula: C4H8Br2
InChi Key: XZNGUVQDFJHPLU-UHFFFAOYNA-N
SMILES: CC(Br)CCBr
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)